Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Metipranolol | 22664-55-7 | 98.4% | 10 MG
SDP

Catalog No. 5000232318
Encompass_Preferred

Metipranolol is a nonselective, orally active β-adrenergic receptor antagonist used in preclinical research for cardiovascular and ocular indications, including hypertension and glaucoma. It is supplied as solid material and as DMSO solutions, and is provided with specification and recommended storage guidance for both solid and solution forms.

  • Nonselective β-adrenergic receptor antagonist suitable for hypertension and glaucoma research.
  • High purity: 98.36% (as supplied).
  • Molecular formula C17H27NO4; molecular weight 309.4 g/mol.
  • Available as solid packs (including 10 mg) and as 10 mM solutions in DMSO.
  • Solubility in DMSO: 250 mg/mL; ultrasonic agitation recommended.
  • Storage: solid at 4°C protected from light; stock solutions at -80°C (6 months) or -20°C (1 month), protected from light.
  • CAS number 22664-55-7 for identification.

Catalog No. 50-002-32318 Supplier Medchemexpress LLC Supplier No. HY12156710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metipranolol is a nonselective, orally active β-adrenergic receptor antagonist used in preclinical research for cardiovascular and ocular indications, including hypertension and glaucoma. It is supplied as solid material and as DMSO solutions, and is provided with specification and recommended storage guidance for both solid and solution forms.

  • Nonselective β-adrenergic receptor antagonist suitable for hypertension and glaucoma research.
  • High purity: 98.36% (as supplied).
  • Molecular formula C17H27NO4; molecular weight 309.4 g/mol.
  • Available as solid packs (including 10 mg) and as 10 mM solutions in DMSO.
  • Solubility in DMSO: 250 mg/mL; ultrasonic agitation recommended.
  • Storage: solid at 4°C protected from light; stock solutions at -80°C (6 months) or -20°C (1 month), protected from light.
  • CAS number 22664-55-7 for identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.