Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Metipranolol | 22664-55-7 | 98.4% | 100 MG
SDP

Catalog No. 5000230794
Encompass_Preferred

Metipranolol is a nonselective, orally active β-adrenergic receptor antagonist commonly used as a research standard for studies related to hypertension and glaucoma. For laboratory use only; consult supplier documentation for safety and handling.

  • Nonselective β-adrenergic receptor antagonist suitable for in vitro and in vivo research.
  • High-purity analytical standard for reliable experimental results.
  • Molecular weight 309.4 g/mol and formula C17H27NO4 for identification and calculations.
  • Soluble in DMSO; follow solubility guidance for selecting solvents.

Catalog No. 50-002-30794 Supplier Medchemexpress LLC Supplier No. HY121567100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metipranolol is a nonselective, orally active β-adrenergic receptor antagonist commonly used as a research standard for studies related to hypertension and glaucoma. For laboratory use only; consult supplier documentation for safety and handling.

  • Nonselective β-adrenergic receptor antagonist suitable for in vitro and in vivo research.
  • High-purity analytical standard for reliable experimental results.
  • Molecular weight 309.4 g/mol and formula C17H27NO4 for identification and calculations.
  • Soluble in DMSO; follow solubility guidance for selecting solvents.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.