Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Metipranolol | 22664-55-7 | 98.4% | 25 MG
SDP

Catalog No. 5000232270
Encompass_Preferred

Metipranolol is a nonselective, orally active β-adrenergic receptor antagonist used in research on hypertension and glaucoma. It is supplied as a research-grade small-molecule solid with characterized purity, solubility, and recommended storage conditions for laboratory use.

  • Nonselective β-adrenergic receptor antagonist used in hypertension and glaucoma research.
  • CAS number 22664-55-7.
  • Molecular formula C17H27NO4 and molecular weight 309.4 g/mol.
  • High purity (≈98.4%), suitable for research applications.
  • Soluble in DMSO (≈250 mg/mL); may require ultrasonic assistance.
  • Recommended storage: solid at 4°C protected from light; in solution store at -80°C up to 6 months or -20°C up to 1 month.
  • Supplied in a 25 MG research pack size.

Catalog No. 50-002-32270 Supplier Medchemexpress LLC Supplier No. HY12156725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metipranolol is a nonselective, orally active β-adrenergic receptor antagonist used in research on hypertension and glaucoma. It is supplied as a research-grade small-molecule solid with characterized purity, solubility, and recommended storage conditions for laboratory use.

  • Nonselective β-adrenergic receptor antagonist used in hypertension and glaucoma research.
  • CAS number 22664-55-7.
  • Molecular formula C17H27NO4 and molecular weight 309.4 g/mol.
  • High purity (≈98.4%), suitable for research applications.
  • Soluble in DMSO (≈250 mg/mL); may require ultrasonic assistance.
  • Recommended storage: solid at 4°C protected from light; in solution store at -80°C up to 6 months or -20°C up to 1 month.
  • Supplied in a 25 MG research pack size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.