Learn More
Medchemexpress LLC HY-16316 5mg Medchemexpress, Metipranolol hydrochloride CAS:36592-77-5 Purity:>98%
SDP
Catalog No. 502081880
Encompass_Preferred
Medchemexpress, HY-16316 5mg Metipranolol hydrochloride CAS:36592-77-5 Metipranolol hydrochloride is a non-selective β adrenergic receptor blocking agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1880 Supplier Medchemexpress LLC Supplier No. HY163165MG
Medchemexpress, HY-16316 5mg Metipranolol hydrochloride CAS:36592-77-5 Metipranolol hydrochloride is a non-selective β adrenergic receptor blocking agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.