Learn More
Medchemexpress LLC Metipranolol hydrochloride | 36592-77-5 | 345.86 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY16316
Encompass_Preferred
Metipranolol hydrochloride is a non-selective β adrenergic receptor blocking agent. This compound targets the adrenergic receptor and is relevant for research in cardiovascular disease and endocrinology. It is also used systemically for the treatment of arterial hypertension and topically for the management of glaucoma.
  • Non-selective β adrenergic receptor blocking agent
  • Targets adrenergic receptor
  • Relevant for cardiovascular disease research
  • Relevant for endocrinology research
  • Used systemically for arterial hypertension
  • Used topically for glaucoma management
Catalog No. 50-003-95025