Learn More
Medchemexpress LLC Metipranolol hydrochloride | 36592-77-5 | MFCD04034693 | 99.9% | 345.86 | C17H28ClNO4 | 10 MG
SDP
Catalog No. 5000288813
Encompass_Preferred
Metipranolol hydrochloride is a non-selective β-adrenergic receptor antagonist supplied as a white to off-white solid for research use. It is commonly used in pharmacology and ophthalmology research related to arterial hypertension and glaucoma. Reported analytical purity is 99.92% and the compound has molecular weight 345.86 (C17H28ClNO4).
  • Non-selective β-adrenergic antagonist suitable for hypertension and glaucoma research.
  • High reported purity for consistent experimental results.
  • Available in small-scale packages for laboratory studies.
  • Solid form simplifies storage and handling; stable under recommended conditions.
Catalog No. 50-002-88813 Supplier Medchemexpress LLC Supplier No. HY1631610MG
Metipranolol hydrochloride is a non-selective β-adrenergic receptor antagonist supplied as a white to off-white solid for research use. It is commonly used in pharmacology and ophthalmology research related to arterial hypertension and glaucoma. Reported analytical purity is 99.92% and the compound has molecular weight 345.86 (C17H28ClNO4).
  • Non-selective β-adrenergic antagonist suitable for hypertension and glaucoma research.
  • High reported purity for consistent experimental results.
  • Available in small-scale packages for laboratory studies.
  • Solid form simplifies storage and handling; stable under recommended conditions.