Learn More
Medchemexpress LLC Metixene hydrochloride hydrate | 7081-40-5 | 99.9% | 25 MG
SDP
Catalog No. 5000234108
Encompass_Preferred
Metixene hydrochloride hydrate is an anticholinergic, antiparkinsonian research standard used as a muscarinic acetylcholine receptor antagonist in pharmacology and neuroscience studies. It is supplied as the hydrochloride hydrate salt for improved stability and solubility and is typically provided in small quantities for laboratory research.
  • Acts as a muscarinic acetylcholine receptor (mAChR) antagonist for receptor binding and functional assays.
  • High purity suitable for analytical and biochemical applications (≈99.9%).
  • Molecular weight 363.94 g/mol and formula C20H26ClNOS.
  • Available in small package sizes such as 25 MG for laboratory use.
  • Provided as a hydrochloride hydrate salt to aid solubility in common solvents.
Catalog No. 50-002-34108 Supplier Medchemexpress LLC Supplier No. HY120081A25MG
Metixene hydrochloride hydrate is an anticholinergic, antiparkinsonian research standard used as a muscarinic acetylcholine receptor antagonist in pharmacology and neuroscience studies. It is supplied as the hydrochloride hydrate salt for improved stability and solubility and is typically provided in small quantities for laboratory research.
  • Acts as a muscarinic acetylcholine receptor (mAChR) antagonist for receptor binding and functional assays.
  • High purity suitable for analytical and biochemical applications (≈99.9%).
  • Molecular weight 363.94 g/mol and formula C20H26ClNOS.
  • Available in small package sizes such as 25 MG for laboratory use.
  • Provided as a hydrochloride hydrate salt to aid solubility in common solvents.