Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MetK protein, Shigella sonnei (His) | 50UG
SDP

Catalog No. 5000176804
Encompass_Preferred

Recombinant metK (S-adenosylmethionine synthase) from Shigella sonnei, expressed in E. coli and supplied as an N-terminal His-tagged purified protein in a filtered formulation. The protein is provided in a HEPES-based buffer with glycerol and DTT, is endotoxin-tested, and is intended for biochemical and structural research; store frozen and avoid repeated freeze-thaw cycles.

  • Recombinant metK from Shigella sonnei, His-tagged.
  • Expressed in E. coli for high yield.
  • Supplied as a 0.22 μm filtered solution in 50 mM HEPES, pH 7.5, 200 mM NaCl, 20% glycerol, 1 mM DTT.
  • Endotoxin level <1 EU/μg, determined by LAL method.
  • Molecular weight ~44 kDa by SDS-PAGE under reducing conditions.
  • Stable when stored at -80°C; ship on dry ice and avoid repeated freeze-thaw cycles.
  • Available in small research sizes suitable for biochemical assays.

Catalog No. 50-001-76804 Supplier Medchemexpress LLC Supplier No. HYP70198350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant metK (S-adenosylmethionine synthase) from Shigella sonnei, expressed in E. coli and supplied as an N-terminal His-tagged purified protein in a filtered formulation. The protein is provided in a HEPES-based buffer with glycerol and DTT, is endotoxin-tested, and is intended for biochemical and structural research; store frozen and avoid repeated freeze-thaw cycles.

  • Recombinant metK from Shigella sonnei, His-tagged.
  • Expressed in E. coli for high yield.
  • Supplied as a 0.22 μm filtered solution in 50 mM HEPES, pH 7.5, 200 mM NaCl, 20% glycerol, 1 mM DTT.
  • Endotoxin level <1 EU/μg, determined by LAL method.
  • Molecular weight ~44 kDa by SDS-PAGE under reducing conditions.
  • Stable when stored at -80°C; ship on dry ice and avoid repeated freeze-thaw cycles.
  • Available in small research sizes suitable for biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.