Learn More
MSC Metalloid Industrial Cleaner, 5 gal., Floor, pH 11, 1 ea
Catalog No. 502820058
Encompass
Metkleen 2140 is a heavy-duty floor and all-purpose maintenance cleaner. This high detergency, hard surface cleaner simultaneously cleans and deodorizes heavily soiled surfaces, with unique wetting properties to enhance detergency and soil penetration. It effectively removes tough oils, greases, and shop soils.
  • Leaves floors clean and dry
  • Hard water stable
  • Multifunctional - cleans and deodorizes
  • Cleans well in cold water
  • Economical - highly concentrated
  • Safe on ferrous and non-ferrous metals
  • Safe on plastic, glass, ceramics, and linoleum
  • Removes tire tracks, grease, carbon, and other soils
Catalog No. 50-282-0058 Supplier MSC Supplier No. 47022496
Metkleen 2140 is a heavy-duty floor and all-purpose maintenance cleaner. This high detergency, hard surface cleaner simultaneously cleans and deodorizes heavily soiled surfaces, with unique wetting properties to enhance detergency and soil penetration. It effectively removes tough oils, greases, and shop soils.
  • Leaves floors clean and dry
  • Hard water stable
  • Multifunctional - cleans and deodorizes
  • Cleans well in cold water
  • Economical - highly concentrated
  • Safe on ferrous and non-ferrous metals
  • Safe on plastic, glass, ceramics, and linoleum
  • Removes tire tracks, grease, carbon, and other soils