Learn More
Cayman Chemical MEtL314 Complex MoncLNal 200ug
SDP
Catalog No. 502990578
Encompass_Preferred
Immunogen: Recombinant human METTL3/14 complex • Host: Mouse • Species Reactivity: (+) Human • Applications: ELISA, WB • MW = 64 and 52 kDa for METTL3 and METTL14, respectively
Catalog No. 50-299-0578 Supplier Cayman Chemical Supplier No. 29313200 UG
Add to Cart
Add to Cart
Immunogen: Recombinant human METTL3/14 complex • Host: Mouse • Species Reactivity: (+) Human • Applications: ELISA, WB • MW = 64 and 52 kDa for METTL3 and METTL14, respectively