Learn More
Electron Microscopy Sciences MetaliTAG™ Cryo Label 2 x 1", Blue on 1" core
SDP
Catalog No. 501930335
Encompass_Preferred
Cryogenic thermal transfer labels for the identification of metal racks, stainless steel cryogenic racks, aluminium cassettes, X-ray film cassettes, etc. Ideal for long-term storage and bio-preservation in liquid or vapor phase liquid nitrogen and extra-low temperature freezers. Labels can be printed with most brands of thermal transfer printers. Temperature range from -196 deg C to +100 deg C.
Size: 2 x 1"
Color: Blue
Core: 1"
Labels across: 1
Perforated: Yes
Catalog No. 50-193-0335 Supplier Electron Microscopy Sciences Supplier No. 7742409BL
Cryogenic thermal transfer labels for the identification of metal racks, stainless steel cryogenic racks, aluminium cassettes, X-ray film cassettes, etc. Ideal for long-term storage and bio-preservation in liquid or vapor phase liquid nitrogen and extra-low temperature freezers. Labels can be printed with most brands of thermal transfer printers. Temperature range from -196 deg C to +100 deg C.
Size: 2 x 1"
Color: Blue
Core: 1"
Labels across: 1
Perforated: Yes