Learn More
Apexbio Technology LLC Metoclopramide 364-62-5 100mg
SDP
Catalog No. 502240571
Encompass_Preferred
Metoclopramide (CAS 364-62-5) is a small molecule that primarily acts as an antagonist at dopamine D2 receptors and also exhibits mixed activity as a 5-HT3 receptor antagonist and 5-HT4 receptor agonist Through inhibition of D2 dopaminergic signaling it enhances gastrointestinal motility and exerts antiemetic effects Metoclopramide has been extensively investigated for applications in managing gastrointestinal disorders such as gastroparesis and gastroesophageal reflux disease as well as for alleviating nausea and vomiting including in pregnant women Research supports the safety profile of this compound for fetal outcomes when administered during pregnancy reinforcing its utility in reproductive and gastrointestinal pharmacology studies
This item is not returnable. View return policy
Catalog No. 50-224-0571 Supplier Apexbio Technology LLC Supplier No. A3599100
Metoclopramide (CAS 364-62-5) is a small molecule that primarily acts as an antagonist at dopamine D2 receptors and also exhibits mixed activity as a 5-HT3 receptor antagonist and 5-HT4 receptor agonist Through inhibition of D2 dopaminergic signaling it enhances gastrointestinal motility and exerts antiemetic effects Metoclopramide has been extensively investigated for applications in managing gastrointestinal disorders such as gastroparesis and gastroesophageal reflux disease as well as for alleviating nausea and vomiting including in pregnant women Research supports the safety profile of this compound for fetal outcomes when administered during pregnancy reinforcing its utility in reproductive and gastrointestinal pharmacology studies