Learn More
Cayman Chemical MetoclopramIde-d3 1mg
SDP
Catalog No. 502940363
Encompass_Preferred
An internal standard for the quantification of metoclopramide by GC- or LC-MS
Catalog No. 50-294-0363 Supplier Cayman Chemical Supplier No. 288851 MG
An internal standard for the quantification of metoclopramide by GC- or LC-MS