Learn More
Medchemexpress LLC Metoclopramide hydrochloride hydrate | 54143-57-6 | 99.96% | 354.27 | 500 MG
SDP
Catalog No. 5000384577
Encompass_Preferred
Metoclopramide hydrochloride hydrate is a potent antagonist of the 5-HT3 and dopamine D2 receptors, with IC50 values of 308 nM and 483 nM, respectively. It is used in research related to nausea, vomiting, gastro-oesophageal reflux, and gastroparesis. This product is for research use only and is not sold to patients.
  • Potent antagonist of 5-HT3 and dopamine D2 receptors.
  • Can be used for research on nausea and vomiting, gastro-oesophageal reflux, and gastroparesis.
Catalog No. 50-003-84577 Supplier Medchemexpress LLC Supplier No. HY17382A500MG
Metoclopramide hydrochloride hydrate is a potent antagonist of the 5-HT3 and dopamine D2 receptors, with IC50 values of 308 nM and 483 nM, respectively. It is used in research related to nausea, vomiting, gastro-oesophageal reflux, and gastroparesis. This product is for research use only and is not sold to patients.
  • Potent antagonist of 5-HT3 and dopamine D2 receptors.
  • Can be used for research on nausea and vomiting, gastro-oesophageal reflux, and gastroparesis.