Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Metolachlor-d6 | 1219803-97-0 | 98.6% | 289.83 g·mol⁻1 | C15H16D6ClNO2 | 10 MG
SDP

Catalog No. 5000297256
Encompass_Preferred

Metolachlor-d6 is the deuterium-labeled analogue of metolachlor, provided as an isotope-labeled reference standard for analytical and tracer applications. It is intended for use as an internal standard in mass spectrometry and related analytical workflows.

  • Deuterium-labeled internal standard for LC-MS/MS quantitation.
  • Chemical purity 98.58% (HPLC).
  • Molecular weight 289.83 g·mol⁻1; CAS 1219803-97-0.
  • Supplied as a 10 MG solid; store at -20°C for long-term stability.
  • Suitable as a tracer in ADME and analytical method development.

Catalog No. 50-002-97256 Supplier Medchemexpress LLC Supplier No. HYB1871S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metolachlor-d6 is the deuterium-labeled analogue of metolachlor, provided as an isotope-labeled reference standard for analytical and tracer applications. It is intended for use as an internal standard in mass spectrometry and related analytical workflows.

  • Deuterium-labeled internal standard for LC-MS/MS quantitation.
  • Chemical purity 98.58% (HPLC).
  • Molecular weight 289.83 g·mol⁻1; CAS 1219803-97-0.
  • Supplied as a 10 MG solid; store at -20°C for long-term stability.
  • Suitable as a tracer in ADME and analytical method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.