Learn More
Medchemexpress LLC Metolachlor-d6 | 1219803-97-0 | 99.6% | 1 MG
SDP
Catalog No. 5000412278
Encompass_Preferred
Metolachlor-d6 is the deuterium-labeled form of Metolachlor. Metolachlor is a pre-emergent, selective, chloroacetanilide herbicide used to control various annual grass and broadleaf weeds in corn and other crops. It is a chiral herbicide consisting of four stereoisomers.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12278 Supplier Medchemexpress LLC Supplier No. HYB1871S1MG
Metolachlor-d6 is the deuterium-labeled form of Metolachlor. Metolachlor is a pre-emergent, selective, chloroacetanilide herbicide used to control various annual grass and broadleaf weeds in corn and other crops. It is a chiral herbicide consisting of four stereoisomers.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.