Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Metolazone-d7 | 2714484-71-4 | 99.9% | 5 MG
SDP

Catalog No. 5000405755
Encompass_Preferred

Metolazone-d7 is deuterium-labeled Metolazone, primarily used to treat congestive heart failure and high blood pressure. Stable heavy isotopes have been incorporated into this drug molecule for use as tracers in drug development, with deuteration potentially affecting pharmacokinetic and metabolic profiles.

  • Molecular weight: 372.88
  • Formula: C16H9D7ClN3O3S
  • Unlabeled CAS: 17560-51-9
  • Appearance: Solid
  • Color: White to off-white
  • Shipped at room temperature (continental US)
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Solubility in DMSO: ≥ 100 mg/mL
  • Solubility in H2O: 0.67 mg/mL (requires ultrasonic)
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-05755 Supplier Medchemexpress LLC Supplier No. HYB0209S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metolazone-d7 is deuterium-labeled Metolazone, primarily used to treat congestive heart failure and high blood pressure. Stable heavy isotopes have been incorporated into this drug molecule for use as tracers in drug development, with deuteration potentially affecting pharmacokinetic and metabolic profiles.

  • Molecular weight: 372.88
  • Formula: C16H9D7ClN3O3S
  • Unlabeled CAS: 17560-51-9
  • Appearance: Solid
  • Color: White to off-white
  • Shipped at room temperature (continental US)
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Solubility in DMSO: ≥ 100 mg/mL
  • Solubility in H2O: 0.67 mg/mL (requires ultrasonic)
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.