Learn More
Medchemexpress LLC Metolazone-d7 | 2714484-71-4 | 99.9% | 500 UG
SDP
Catalog No. 5000405754
Encompass_Preferred
Metolazone-d7 is deuterium labeled Metolazone, primarily used to treat congestive heart failure and high blood pressure. Deuteration, the incorporation of stable heavy isotopes into drug molecules, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05754 Supplier Medchemexpress LLC Supplier No. HYB0209S500UG
Metolazone-d7 is deuterium labeled Metolazone, primarily used to treat congestive heart failure and high blood pressure. Deuteration, the incorporation of stable heavy isotopes into drug molecules, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS