Learn More
U.S. Pharmacopeia Metomidate Hydrochloride, 35944-74-2, MFCD01721715, 50mg
Catalog No. NC1426394
Encompass
Molecular formula C13H14N2O2.HCl, Molecular weight 266.72, Melting point/freezing point 329 - 338 °F (165 - 170 °C) (decomposes), Initial boiling point 714.92 °F (379.4 °C) at 760 mm Hg, Synonyms: 1-(1-Phenethyl)-imidazole-5-carboxylic acid, methyl ester, hydrochloride
Catalog No. NC1426394 Supplier U.S. Pharmacopeia Supplier No. 1268772
Molecular formula C13H14N2O2.HCl, Molecular weight 266.72, Melting point/freezing point 329 - 338 °F (165 - 170 °C) (decomposes), Initial boiling point 714.92 °F (379.4 °C) at 760 mm Hg, Synonyms: 1-(1-Phenethyl)-imidazole-5-carboxylic acid, methyl ester, hydrochloride