Learn More
Beckman Coulter Met One HHPC 2+ Handheld Particle Counter 2 Channel 5um 1 EA
Catalog No. NC0350746 Shop All Beckman Coulter Products
Encompass
Laser-based optical airborne particle counter with 0.5 to 5.0 UM size range in 2 channels, portable battery-powered design with LCD display, data logging & USB connectivity for real-time air quality monitoring & detecting sources of contamination in cleanrooms, hospitals & facilities.
Catalog No. NC0350746 Supplier Beckman Coulter Supplier No. 208931003
Laser-based optical airborne particle counter with 0.5 to 5.0 UM size range in 2 channels, portable battery-powered design with LCD display, data logging & USB connectivity for real-time air quality monitoring & detecting sources of contamination in cleanrooms, hospitals & facilities.