Learn More
Medchemexpress LLC Metoprolol | 51384-51-1 | 99.98% | 267.36 | 5 MG
SDP
Catalog No. 5000387030
Encompass_Preferred
Metoprolol (Standard) is the analytical standard of Metoprolol. This product is intended for research and analytical applications. Metoprolol is an orally active, selective β1-adrenoceptor antagonist. It shows anti-inflammation, antitumor and anti-angiogenic properties.
  • Analytical standard for Metoprolol.
  • Intended for research and analytical applications.
  • Orally active, selective β1-adrenoceptor antagonist.
  • Shows anti-inflammation, antitumor and anti-angiogenic properties.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-003-87030 Supplier Medchemexpress LLC Supplier No. HY17503R5MG
Metoprolol (Standard) is the analytical standard of Metoprolol. This product is intended for research and analytical applications. Metoprolol is an orally active, selective β1-adrenoceptor antagonist. It shows anti-inflammation, antitumor and anti-angiogenic properties.
  • Analytical standard for Metoprolol.
  • Intended for research and analytical applications.
  • Orally active, selective β1-adrenoceptor antagonist.
  • Shows anti-inflammation, antitumor and anti-angiogenic properties.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.