Learn More
Medchemexpress LLC Metoprolol | 51384-51-1 | 99.99% | 267.36 | 100 MG
SDP
Catalog No. 5000386756
Encompass_Preferred
Metoprolol is an orally active, selective β1-adrenoceptor antagonist. It exhibits anti-inflammatory, antitumor, and anti-angiogenic properties.
  • Orally active.
  • Selective β1-adrenoceptor antagonist.
  • Shows anti-inflammation, antitumor, and anti-angiogenic properties.
  • Cytotoxic effect on U937 and MOLT-4 cells dose and time dependently in vitro.
  • Reduces proinflammatory cytokines and atherosclerosis in ApoE-/- mice in vivo.
  • Shows anti-inflammation and anti-virus effects in a murine model with coxsackievirus B3-induced viral myocarditis in vivo.
  • Significantly decreased activated caspase-9 protein expression and inhibits myocardial apoptosis in coronary microembolization (CME) rats in vivo.
Catalog No. 50-003-86756 Supplier Medchemexpress LLC Supplier No. HY17503100MG
Metoprolol is an orally active, selective β1-adrenoceptor antagonist. It exhibits anti-inflammatory, antitumor, and anti-angiogenic properties.
  • Orally active.
  • Selective β1-adrenoceptor antagonist.
  • Shows anti-inflammation, antitumor, and anti-angiogenic properties.
  • Cytotoxic effect on U937 and MOLT-4 cells dose and time dependently in vitro.
  • Reduces proinflammatory cytokines and atherosclerosis in ApoE-/- mice in vivo.
  • Shows anti-inflammation and anti-virus effects in a murine model with coxsackievirus B3-induced viral myocarditis in vivo.
  • Significantly decreased activated caspase-9 protein expression and inhibits myocardial apoptosis in coronary microembolization (CME) rats in vivo.