Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Metoprolol | 51384-51-1 | MFCD00599534 | 100.0% | 267.36 g·mol-1 | C15H25NO3 | 10 MG
SDP

Catalog No. 5000298175
Encompass_Preferred

Metoprolol analytical standard intended for research and analytical applications. Supplied as a 10 mg vial, this reference material supports assay development, method validation, and compound identification; it is intended for laboratory use only and not for human or clinical use.

  • Analytical reference standard for assay development and method validation.
  • High purity (>99.9%) suitable for chromatographic analysis.
  • Molecular formula C15H25NO3; molecular weight 267.36 g·mol-1.
  • Provided in small research packaging for laboratory use only.
  • Not for human or clinical use.

Catalog No. 50-002-98175 Supplier Medchemexpress LLC Supplier No. HY17503R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metoprolol analytical standard intended for research and analytical applications. Supplied as a 10 mg vial, this reference material supports assay development, method validation, and compound identification; it is intended for laboratory use only and not for human or clinical use.

  • Analytical reference standard for assay development and method validation.
  • High purity (>99.9%) suitable for chromatographic analysis.
  • Molecular formula C15H25NO3; molecular weight 267.36 g·mol-1.
  • Provided in small research packaging for laboratory use only.
  • Not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.