Learn More
Medchemexpress LLC Metoprolol-d5 | 959786-79-9 | 96.9% | 272.39 | 1 MG
SDP
Catalog No. 5000386865
Encompass_Preferred
Metoprolol-d5 is the deuterium labeled Metoprolol, an orally active, selective β1-adrenoceptor antagonist. It exhibits anti-inflammation, antitumor, and anti-angiogenic properties. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development, with deuteration specifically impacting pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-86865 Supplier Medchemexpress LLC Supplier No. HY17503S31MG
Metoprolol-d5 is the deuterium labeled Metoprolol, an orally active, selective β1-adrenoceptor antagonist. It exhibits anti-inflammation, antitumor, and anti-angiogenic properties. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development, with deuteration specifically impacting pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.