Learn More
Medchemexpress LLC Metoprolol-d7 hydrochloride | 1219798-61-4 | 99.7% | 310.87 | 5 MG
SDP
Catalog No. 5000386770
Encompass_Preferred
Metoprolol-d7 hydrochloride is a deuterium-labeled form of Metoprolol, an orally active, selective β1-adrenoceptor antagonist. It demonstrates anti-inflammatory, antitumor, and anti-angiogenic properties.
  • Deuterium labeled compound
  • Selective β1-adrenoceptor antagonist
  • Exhibits anti-inflammatory, antitumor, and anti-angiogenic properties
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-86770 Supplier Medchemexpress LLC Supplier No. HY17503AS5MG
Metoprolol-d7 hydrochloride is a deuterium-labeled form of Metoprolol, an orally active, selective β1-adrenoceptor antagonist. It demonstrates anti-inflammatory, antitumor, and anti-angiogenic properties.
  • Deuterium labeled compound
  • Selective β1-adrenoceptor antagonist
  • Exhibits anti-inflammatory, antitumor, and anti-angiogenic properties
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS