Learn More
Medchemexpress LLC Metoprolol (Standard) | 51384-51-1 | 99.99% | 267.36 | 1 G
SDP
Catalog No. 5000386779
Encompass_Preferred
Metoprolol is an orally active, selective β1-adrenoceptor antagonist. It exhibits anti-inflammation, antitumor, and anti-angiogenic properties, making it suitable for various research applications.
  • Acts as a selective β1-adrenoceptor antagonist
  • Exhibits anti-inflammatory effects
  • Demonstrates antitumor activity
  • Shows anti-angiogenic properties
  • Cytotoxic to U937 and MOLT-4 cells in vitro
  • Reduces proinflammatory cytokines and atherosclerosis in ApoE-/- mice
  • Provides anti-inflammatory and antiviral effects in viral myocarditis models
  • Inhibits myocardial apoptosis in coronary microembolization models
Catalog No. 50-003-86779 Supplier Medchemexpress LLC Supplier No. HY175031G
Metoprolol is an orally active, selective β1-adrenoceptor antagonist. It exhibits anti-inflammation, antitumor, and anti-angiogenic properties, making it suitable for various research applications.
  • Acts as a selective β1-adrenoceptor antagonist
  • Exhibits anti-inflammatory effects
  • Demonstrates antitumor activity
  • Shows anti-angiogenic properties
  • Cytotoxic to U937 and MOLT-4 cells in vitro
  • Reduces proinflammatory cytokines and atherosclerosis in ApoE-/- mice
  • Provides anti-inflammatory and antiviral effects in viral myocarditis models
  • Inhibits myocardial apoptosis in coronary microembolization models