Learn More
Apexbio Technology LLC Metoprolol Succinate 98418-47-4 50mg
SDP
Catalog No. 5000092799
Encompass_Preferred
Metoprolol Succinate (CAS 98418-47-4) is an orally active selective 1-adrenergic receptor antagonist By selectively inhibiting 1-receptors in cardiac tissue it modulates myocardial contractility and heart rate resulting in altered hemodynamic parameters In preclinical cardiovascular models this small molecule is utilized to investigate its effects on cardiac function and systemic blood pressure Owing to its receptor selectivity Metoprolol Succinate is commonly employed in studies exploring therapeutic strategies for chronic heart failure and hypertension
Catalog No. 50-000-92799 Supplier Apexbio Technology LLC Supplier No. B602250
Metoprolol Succinate (CAS 98418-47-4) is an orally active selective 1-adrenergic receptor antagonist By selectively inhibiting 1-receptors in cardiac tissue it modulates myocardial contractility and heart rate resulting in altered hemodynamic parameters In preclinical cardiovascular models this small molecule is utilized to investigate its effects on cardiac function and systemic blood pressure Owing to its receptor selectivity Metoprolol Succinate is commonly employed in studies exploring therapeutic strategies for chronic heart failure and hypertension