Learn More
Selleck Chemical LLC Metoprolol Tartrate S1856-50mg
SDP
Catalog No. 503109596
Encompass_Preferred
Metoprolol Tartrate (CGP 2175E) is a selective 1 receptor blocker medication used to treat hypertension and heart failure
Catalog No. 50-310-9596 Supplier Selleck Chemical LLC Supplier No. S185650MG
Metoprolol Tartrate (CGP 2175E) is a selective 1 receptor blocker medication used to treat hypertension and heart failure