Learn More
Medchemexpress LLC Metoprolol tartrate | 56392-17-7 | 100.0% | 342.41 | 200 MG
SDP
Catalog No. 5000386766
Encompass_Preferred
Metoprolol tartrate is an orally active, selective β1-adrenoceptor antagonist. It exhibits anti-inflammation, antitumor, and anti-angiogenic properties. This product is intended for research use only and is not sold to patients.
  • Acts as a selective β1-adrenoceptor antagonist.
  • Exhibits anti-inflammatory effects.
  • Demonstrates antitumor properties.
  • Shows anti-angiogenic effects.
  • Cytotoxic to U937 and MOLT-4 cells.
  • Reduces proinflammatory cytokines.
  • Decreases myocardial virus titers.
  • Inhibits myocardial apoptosis.
Catalog No. 50-003-86766 Supplier Medchemexpress LLC Supplier No. HY17503B200MG
Metoprolol tartrate is an orally active, selective β1-adrenoceptor antagonist. It exhibits anti-inflammation, antitumor, and anti-angiogenic properties. This product is intended for research use only and is not sold to patients.
  • Acts as a selective β1-adrenoceptor antagonist.
  • Exhibits anti-inflammatory effects.
  • Demonstrates antitumor properties.
  • Shows anti-angiogenic effects.
  • Cytotoxic to U937 and MOLT-4 cells.
  • Reduces proinflammatory cytokines.
  • Decreases myocardial virus titers.
  • Inhibits myocardial apoptosis.