Learn More
Medchemexpress LLC Metoprolol tartrate | 56392-17-7 | 99.98% | 342.41 | 50 MG
SDP
Catalog No. 5000386798
Encompass_Preferred
Metoprolol tartrate is a selective β1-adrenoceptor antagonist that is orally active. It has demonstrated anti-inflammatory, antitumor, and anti-angiogenic properties. This compound is intended for research use only and exhibits dose and time-dependent cytotoxic effects on U937 and MOLT-4 cells in vitro, and reduces proinflammatory cytokines and atherosclerosis in ApoE-/- Mice in vivo.
  • Selective β1-adrenoceptor antagonist.
  • Orally active.
  • Multi-faceted properties: anti-inflammatory, antitumor, and anti-angiogenic effects.
  • Exhibits cytotoxic effects on U937 and MOLT-4 cells in vitro.
  • Reduces proinflammatory cytokines and atherosclerosis in ApoE-/- Mice.
  • Exhibits anti-inflammation and anti-virus effects in a murine model with coxsackievirus B3-induced viral myocarditis.
  • Inhibits myocardial apoptosis in coronary microembolization (CME) rats.
Catalog No. 50-003-86798 Supplier Medchemexpress LLC Supplier No. HY17503B50MG
Metoprolol tartrate is a selective β1-adrenoceptor antagonist that is orally active. It has demonstrated anti-inflammatory, antitumor, and anti-angiogenic properties. This compound is intended for research use only and exhibits dose and time-dependent cytotoxic effects on U937 and MOLT-4 cells in vitro, and reduces proinflammatory cytokines and atherosclerosis in ApoE-/- Mice in vivo.
  • Selective β1-adrenoceptor antagonist.
  • Orally active.
  • Multi-faceted properties: anti-inflammatory, antitumor, and anti-angiogenic effects.
  • Exhibits cytotoxic effects on U937 and MOLT-4 cells in vitro.
  • Reduces proinflammatory cytokines and atherosclerosis in ApoE-/- Mice.
  • Exhibits anti-inflammation and anti-virus effects in a murine model with coxsackievirus B3-induced viral myocarditis.
  • Inhibits myocardial apoptosis in coronary microembolization (CME) rats.