Learn More
Medchemexpress LLC Metoprolol tartrate | 56392-17-7 | MFCD00056257 | 100.0% | 342.41 g·mol⁻¹ | C19H31NO9 | 10 MG
SDP
Catalog No. 5000298532
Encompass_Preferred
Metoprolol tartrate is the tartrate salt of metoprolol, a selective β1-adrenoceptor antagonist used in research to study β1 receptor pharmacology and cardiovascular effects. It is supplied as a purified chemical reference for in vitro and preclinical studies, and has been reported to exhibit anti-inflammatory, antitumor, and anti-angiogenic activities.
  • Selective β1-adrenoceptor antagonist for receptor pharmacology studies.
  • Documented anti-inflammatory, antitumor, and anti-angiogenic properties.
  • High purity suitable for analytical and experimental use.
  • Available in small research pack sizes for assay development.
  • Suitable as a chemical reference standard in in vitro and preclinical assays.
  • Solid form, compatible with standard laboratory handling procedures.
Catalog No. 50-002-98532 Supplier Medchemexpress LLC Supplier No. HY17503B10MG
Metoprolol tartrate is the tartrate salt of metoprolol, a selective β1-adrenoceptor antagonist used in research to study β1 receptor pharmacology and cardiovascular effects. It is supplied as a purified chemical reference for in vitro and preclinical studies, and has been reported to exhibit anti-inflammatory, antitumor, and anti-angiogenic activities.
  • Selective β1-adrenoceptor antagonist for receptor pharmacology studies.
  • Documented anti-inflammatory, antitumor, and anti-angiogenic properties.
  • High purity suitable for analytical and experimental use.
  • Available in small research pack sizes for assay development.
  • Suitable as a chemical reference standard in in vitro and preclinical assays.
  • Solid form, compatible with standard laboratory handling procedures.