Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metoprolol Tartrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014635 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4635 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4635 Supplier MilliporeSigma Supelco Supplier No. PHR10761G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Metoprolol Tartrate is a β-blocking drug, showing antihypertensive and anti-ischemic effects. It is widely used in the treatment of hypertension and helps in controlling ventricular response in patients with atrial fibrillation.

TRUSTED_SUSTAINABILITY

Specifications

CAS 56392-17-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula (C15H25 NO3)2 · C4H6O6
MDL Number MFCD00056257
Synonym (+/-)-Metoprolol (+)-tartrate salt; (+/-)1-(Isopropylamino)-3-[p-(beta-methoxyethyl)phenoxy]-2-propanol (+)-tartrate salt; Lopressor
Type Certified Reference Material
Quantity 1 g
Formula Weight 684.81
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.