Learn More
Cayman Chemical Metoprolol (succinate), Purity: ≥98%, CAS: 98418-47-4, 50 mg
SDP
Catalog No. 502969123
Encompass_Preferred
A β1-AR antagonist (Ki = 47 nM in CHO cells expressing the human receptor); selective for β1- over β2- and β3-ARs (Kis = 2,730 and >10,000 nM, respectively, in CHO cells expressing the human receptors); reduces GTP-induced AC activity in CHO cells expressing β1- and β2-ARs at 4.7 µM; reduces the pulse rate in isolated rat atria in a concentration-dependent manner; reduces increased heart rate and MAP at 30 mg/kg in a rat model of systolic hypertension induced by a fructose-rich diet
Catalog No. 50-296-9123 Supplier Cayman Chemical Supplier No. 1542950 MG
A β1-AR antagonist (Ki = 47 nM in CHO cells expressing the human receptor); selective for β1- over β2- and β3-ARs (Kis = 2,730 and >10,000 nM, respectively, in CHO cells expressing the human receptors); reduces GTP-induced AC activity in CHO cells expressing β1- and β2-ARs at 4.7 µM; reduces the pulse rate in isolated rat atria in a concentration-dependent manner; reduces increased heart rate and MAP at 30 mg/kg in a rat model of systolic hypertension induced by a fructose-rich diet