Learn More
Bellco Glass, Inc. Metric 3-Port Tube Assembly For 18mm Port
SDP
Catalog No. 501946422
Encompass_Preferred
3-port tube assembly. MADE IN THE USAThree (3) .125" od tubes welded in to 18mm plug portConsist of:3-hole port plug(3) .125 tubes#115 silicone o-ringSST knurled cap nutSST washer
Catalog No. 50-194-6422 Supplier Bellco Glass, Inc. Supplier No. 790900006
3-port tube assembly. MADE IN THE USAThree (3) .125" od tubes welded in to 18mm plug portConsist of:3-hole port plug(3) .125 tubes#115 silicone o-ringSST knurled cap nutSST washer