Learn More
Bellco Glass, Inc. Metric Bioreactor Stand, 15L
SDP
Catalog No. 501946484
Encompass_Preferred
Vessel stand for water jacketed metric bioreactor vessel has unique split ring design to fit around jacket and hose barbs. Manufactured from aluminum and stainless steel for durability and ease of cleaning. Bottom ring insures stability of vessel during operation. MADE IN THE USA
Catalog No. 50-194-6484 Supplier Bellco Glass, Inc. Supplier No. 790915088
Vessel stand for water jacketed metric bioreactor vessel has unique split ring design to fit around jacket and hose barbs. Manufactured from aluminum and stainless steel for durability and ease of cleaning. Bottom ring insures stability of vessel during operation. MADE IN THE USA