Learn More
Bellco Glass, Inc. Metric Bioreactor Stand, 3L
SDP
Catalog No. 501946430
Encompass_Preferred
Metric Bioreactor Stand Vessel stand for water jacketed metric bioreactorvessel has unique split ring design to fit around jacket and hose barbs.Manufactured from aluminum and stainless steel for durability and ease of cleaning. Bottom ring insures stability of vessel during operation. 3L, size: 10 7/8”. MADE IN THE USA
Catalog No. 50-194-6430 Supplier Bellco Glass, Inc. Supplier No. 790900019
Metric Bioreactor Stand Vessel stand for water jacketed metric bioreactorvessel has unique split ring design to fit around jacket and hose barbs.Manufactured from aluminum and stainless steel for durability and ease of cleaning. Bottom ring insures stability of vessel during operation. 3L, size: 10 7/8”. MADE IN THE USA