Learn More
Bellco Glass, Inc. Metric Bioreactor Stand, 7L
SDP
Catalog No. 501946449
Encompass_Preferred
Vessel stand for 3L water jacketed metric bioreactor vessel has unique split ring design to fit around jacket and hose barbs. Manufactured from aluminum and stainless steel for durability and ease of cleaning. Bottom ring insures stability of vessel during operation.. MADE IN THE USA
Catalog No. 50-194-6449 Supplier Bellco Glass, Inc. Supplier No. 790900043
Vessel stand for 3L water jacketed metric bioreactor vessel has unique split ring design to fit around jacket and hose barbs. Manufactured from aluminum and stainless steel for durability and ease of cleaning. Bottom ring insures stability of vessel during operation.. MADE IN THE USA