Learn More
Bellco Glass, Inc. Metric Condenser Assembly , 3L
SDP
Catalog No. 501946423
Encompass_Preferred
Stainless steel condenser mounted externally has internal baffle for efficient condensation of vapor into liquid for return to bioreactor vessel. Fits 18mm port on 3L metric headplate. 316L Stainless Steel Construction Ra 32 Finish Or Better. MADE IN THE USA
Catalog No. 50-194-6423 Supplier Bellco Glass, Inc. Supplier No. 790900008
Stainless steel condenser mounted externally has internal baffle for efficient condensation of vapor into liquid for return to bioreactor vessel. Fits 18mm port on 3L metric headplate. 316L Stainless Steel Construction Ra 32 Finish Or Better. MADE IN THE USA