Learn More
Bellco Glass, Inc. Metric Condenser Assembly , 7L
SDP
Catalog No. 501946453
Encompass_Preferred
Stainless steal condenser mounted externally has internal baffle for efficient condensation of vapor into liquid for return to bioreactor vessel. Fits 18mm port on 7L metric headplate. 316L. Condenser assembly, 7L. MADE IN THE USA
Catalog No. 50-194-6453 Supplier Bellco Glass, Inc. Supplier No. 790900047
Stainless steal condenser mounted externally has internal baffle for efficient condensation of vapor into liquid for return to bioreactor vessel. Fits 18mm port on 7L metric headplate. 316L. Condenser assembly, 7L. MADE IN THE USA