Learn More
Bellco Glass, Inc. Metric Drive Stem Assembly , 3L for Mag motor
SDP
Catalog No. 501946419
Encompass_Preferred
Drive stem assembly with 3/4" OD shaft for use with 3L marine and pitched blade impellers. Fits into 27 mm center port of 3L metric headplate. Multiple impellers may be assembled onto shaft as required.. MADE IN THE USA
Catalog No. 50-194-6419 Supplier Bellco Glass, Inc. Supplier No. 790900003
Add to Cart
Add to Cart
Drive stem assembly with 3/4" OD shaft for use with 3L marine and pitched blade impellers. Fits into 27 mm center port of 3L metric headplate. Multiple impellers may be assembled onto shaft as required.. MADE IN THE USA