Learn More
Bellco Glass, Inc. Metric Drive Stem Assembly , 7L for mag motor
SDP
Catalog No. 501946463
Encompass_Preferred
Drive stem assembly with 3/4”OD shaft for use with 7L marine and pitched blade impellers. Fits into 27 mm center port of 7L metric headplates. Multiple impellers may be assembled onto shaft as required.. MADE IN THE USA
Catalog No. 50-194-6463 Supplier Bellco Glass, Inc. Supplier No. 790907004
Add to Cart
Add to Cart
Drive stem assembly with 3/4”OD shaft for use with 7L marine and pitched blade impellers. Fits into 27 mm center port of 7L metric headplates. Multiple impellers may be assembled onto shaft as required.. MADE IN THE USA