Learn More
Bellco Glass, Inc. Metric Head Plate, 15L 20 Holes
SDP
Catalog No. 501946474
Encompass_Preferred
Material: 316l stainless steelTop finish: 400 mirror polishBottom finish: ra32 electropolishDimensions: 12" od x 3/8" thick# of holes: 20Hole design:10mm hole: 1218mm hole: 8Supply with a555-376 - #448 o-ring. MADE IN THE USA
Catalog No. 50-194-6474 Supplier Bellco Glass, Inc. Supplier No. 790915000
Material: 316l stainless steelTop finish: 400 mirror polishBottom finish: ra32 electropolishDimensions: 12" od x 3/8" thick# of holes: 20Hole design:10mm hole: 1218mm hole: 8Supply with a555-376 - #448 o-ring. MADE IN THE USA