Learn More
Bellco Glass, Inc. Metric Head Plate, 3L
SDP
Catalog No. 501946503
Encompass_Preferred
Material: 316l stainless steelTop finish: 400 mirror polishBottom finish: ra32 electropolishDimensions: 8x3/8”# of holes: 20Hole design:10mm hole: 1218mm hole: 8Supply with a555-376 – #448 o-ring. MADE IN THE USA
Catalog No. 50-194-6503 Supplier Bellco Glass, Inc. Supplier No. 790930000
Material: 316l stainless steelTop finish: 400 mirror polishBottom finish: ra32 electropolishDimensions: 8x3/8”# of holes: 20Hole design:10mm hole: 1218mm hole: 8Supply with a555-376 – #448 o-ring. MADE IN THE USA