Learn More
Bellco Glass, Inc. Metric Head Plate, 7L
SDP
Catalog No. 501946504
Encompass_Preferred
Material: 316l stainless steelTop finish: 400 mirror polishBottom finish: ra32 electropolishDimensions: 9¾x 3/8”# of holes: 20Hole design:10mm hole: 1218mm hole: 8Supply with a555-376 – #448 o-ring. MADE IN THE USA
Catalog No. 50-194-6504 Supplier Bellco Glass, Inc. Supplier No. 790970000
Material: 316l stainless steelTop finish: 400 mirror polishBottom finish: ra32 electropolishDimensions: 9¾x 3/8”# of holes: 20Hole design:10mm hole: 1218mm hole: 8Supply with a555-376 – #448 o-ring. MADE IN THE USA