Learn More
MSC BluShield Pressure Washer Gun, Metric, 3000 PSI, Brass
Catalog No. 502837941
Encompass
This pressure washer gun is designed for use with metric pressure washers and accessories, featuring a male metric inlet for connecting to hoses with female metric fittings and a female metric outlet for direct linkage to metric wands.
  • Trigger lock helps prevent accidental spraying
  • Designed for use with metric wands and accessories
  • Constructed from durable brass
  • Withstands a minimum pressure of 3000 psi
Catalog No. 50-283-7941 Supplier MSC Supplier No. 63959332
This pressure washer gun is designed for use with metric pressure washers and accessories, featuring a male metric inlet for connecting to hoses with female metric fittings and a female metric outlet for direct linkage to metric wands.
  • Trigger lock helps prevent accidental spraying
  • Designed for use with metric wands and accessories
  • Constructed from durable brass
  • Withstands a minimum pressure of 3000 psi