Learn More
Grainger Metric Rim Hole Valve
Catalog No. 502750750
Encompass_Preferred
The HALTEC Metric Rim Hole Valve is a metric rim hole valve with a brass body and a stainless steel stem. It has a maximum pressure rating of 1034 kPa and temperature range of -29 to 93°C. The valve has female NPT thread connections and is suitable for air, water, oil, and inert gas applications. The valve features a chrome plated brass handle with a molded PVC grip.
Catalog No. 50-275-0750 Supplier Grainger Supplier No. 140U84
The HALTEC Metric Rim Hole Valve is a metric rim hole valve with a brass body and a stainless steel stem. It has a maximum pressure rating of 1034 kPa and temperature range of -29 to 93°C. The valve has female NPT thread connections and is suitable for air, water, oil, and inert gas applications. The valve features a chrome plated brass handle with a molded PVC grip.