Learn More
MSC Balax Thread Plug Gage, M6 x 1.0, 6H, Single End, 1 ea
Catalog No. 5000513271
Encompass
This single end, go plug thread gage is designed for precise measurement of M6x1 metric threads. Constructed from high speed tool steel with a bright finish, it offers durable performance for inspecting thread accuracy in metalworking applications.
  • Thread size: M6x1
  • Class of fit: 6H
  • Construction: High speed tool steel
  • Hardness: 63 to 65 Rockwell
  • Overall length: 1-3/4 in.
  • Thread length: 1/2 in.
  • Handle size: #1 taperlock
Catalog No. 50-005-13271 Supplier MSC Supplier No. 05337605
This single end, go plug thread gage is designed for precise measurement of M6x1 metric threads. Constructed from high speed tool steel with a bright finish, it offers durable performance for inspecting thread accuracy in metalworking applications.
  • Thread size: M6x1
  • Class of fit: 6H
  • Construction: High speed tool steel
  • Hardness: 63 to 65 Rockwell
  • Overall length: 1-3/4 in.
  • Thread length: 1/2 in.
  • Handle size: #1 taperlock