Learn More
IDEX Health & Science Upchurch METRIC THREADED ADAPTER 1/4-28
Catalog No. NC1509094
Encompass_Preferred
Metric Threaded Adapter 1/4-28-Female to M6-Male
Catalog No. NC1509094 Supplier IDEX Health & Science Upchurch Supplier No. P694
Metric Threaded Adapter 1/4-28-Female to M6-Male