Description
- UNITRON's software designed for industrial materials, scientists, metallurgists, laboratory technicians, and research and design engineers who need improved solutions for quality control and product development.
- METRICAL's computer-aided quantitative analysis results strictly follow ISO and ASTM technical standards.
- The software is powerful to handle different image format BMP, TIFF, GIF, JPEG, etc. It supports several of our Excelis™ and Teledyne Lumenera cameras, as well as TWAIN input devices.
- This exceptional software produces as much as 80% more picture points in image window than similar software for metallurgy and materials science.
- Computerized image processing techniques aide to streamline image capture, measurement, and enhancement, while improving the accuracy of your results.
- METRICAL Metallurgical and Materials Analysis Software in accordance with ISO and ASTM standards.
- Includes modules for grain size, porosity, segmentation, decarburization, ect.
- Compatible with Excelis™ 4K, Excelis™ HD, Excelis™ MPX-5C Pro, and Lumenera INFINITY 3 and INFINITY 5 cameras (must order separately). Not compatible with Excelis™ Lite cameras.
- Windows operating system only.
- Compatible with any brand of microscope, although we would prefer you use UNITRON.
Specifications
Specifications
| Type | Microscope Software |
| Autoclavable | Not Autoclavable |
| Sterility | Non-sterile |