Learn More
Medex Supply METRICIDE OPA PLUS SLTN,4/CS
SDP
Catalog No. NC1450610
Encompass
Metrex MetriCide OPA Plus Solution, 1 Gallon Bottle, 4/CsMetriCide OPA Plus Solution provides a broad spectrum of kill. It was tested against viruses representative of the categories required by the FDA, in addition to other viruses that pose a threat or are representative of viruses that can jeopardize the well being of health care professionals. These viruses include: Hepatitis A (AER) and B and the Avian Influenza A. See table below indicating the entire spectrum of activity demonstrated by testing of MetriCide OPA Plus Solution using prescribed test methods. Bactericidal, fungicidal, tuberculocidal, and sporicidal tests all passed in 5 minutes at 25°C.
This item is not returnable. View return policy
Catalog No. NC1450610 Supplier Medex Supply Supplier No. MRX106000
Metrex MetriCide OPA Plus Solution, 1 Gallon Bottle, 4/CsMetriCide OPA Plus Solution provides a broad spectrum of kill. It was tested against viruses representative of the categories required by the FDA, in addition to other viruses that pose a threat or are representative of viruses that can jeopardize the well being of health care professionals. These viruses include: Hepatitis A (AER) and B and the Avian Influenza A. See table below indicating the entire spectrum of activity demonstrated by testing of MetriCide OPA Plus Solution using prescribed test methods. Bactericidal, fungicidal, tuberculocidal, and sporicidal tests all passed in 5 minutes at 25°C.